Learn More
Stat6 (pY641) Mouse anti-Human, PE, Clone: 18/P-STAT6, BD
Mouse Monoclonal Antibody
525.00€ - 1810.00€
Specifications
Antigen | Stat6 (pY641) |
---|---|
Clone | 18/P-STAT6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15806269
|
BD Biosciences
612701 |
50 Tests |
525.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15811629
|
BD Biosciences
562078 |
250 Tests |
1810.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
Interleukin-4 (IL-4), a major immunoregulatory cytokine, is secreted by activated T lymphocytes, basophils, and mast cells and plays an important role in modulating T helper cell lineage development. It induces specific gene expression via the tyrosine phosphorylation of Stat6 at tyrosine 641 (Y641). Stat6, a member of the signal transducers and activators of transcription protein family, mediates signals for IL-4 and, possibly, IL-13. While Stat6 is widely expressed in human tissues, it exhibits elevated expression in peripheral blood lymphocytes, colon, intestine, ovary, prostate, thymus, spleen, kidney, liver, lung, and placenta. Following cytokine receptor ligation, Jak kinases are activated and phosphorylate the cytoplasmic tails of the oligomerized receptors. The SH3:SH2 domain of Stat6 associates with tyrosine-phosphorylated IL-4 receptor and the proximal Jak kinase phosphorylates Stat6 at Y641 on the C-terminal side of the SH2 domain. Stat6 is then released from the receptor, dimerizes, and is thought to contact the basal transcription machinery by binding to p300/CBP. Thus, Stat6 mediates the IL-4 signal and is essential for the proper development of adaptive immunity.
Specifications
Stat6 (pY641) | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG2a | |
Affinity Purified | |
Signal transducer and activator of transcription 6; IL-4 Stat |
18/P-STAT6 | |
Monoclonal | |
Mouse | |
Human | |
Phosphorylated Human Stat6 (pY641) | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.