missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Starter Kit for MethoCult™ H4434 Classic
Description
- The Starter Kit for MethoCult™ H4434 Classic (MethoCult™ GF H4434) is recommended for laboratories in the initial stages of establishing procedures for assessing human hematopoietic progenitor cells using colony-forming unit (CFU) assays
- These products support the growth of clonogenic hematopoietic progenitor cells from human bone marrow, peripheral blood, cord blood, leukapheresis products, and purified progenitor-enriched cells
- H4434 is a complete methylcellulose-based medium that supports the growth of erythroid progenitors (BFU-E and CFU-E), granulocyte-macrophage progenitors (CFU-GM, CFU-G and CFU-M) and multi-potential granulocyte, erythroid, macrophage, megakaryocyte progenitors (CFU-GEMM)
- Each kit contains instructional materials in addition to all reagents and materials necessary to perform 24 duplicate assays
Specifications
Specifications
| Cell Type | Hematopoietic Stem and Progenitor Cells |
| For Use With (Application) | Cell Culture, Colony Assay, Functional Assay |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?