Learn More
Thermo Scientific™ Staphytect Plus™ Latex Agglutination Test
Product Code. 11483949
Change view
Click to view available options
Quantity:
100 Tests/Kit
500 Tests/Kit
Unit Size:
100 tests
500 tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483949 100 Tests/Kit 100 tests
11473949 500 Tests/Kit 500 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11483949 Supplier Thermo Scientific™ Supplier No. DR0850M
 Restricted Item
Estimated Shipment: 26-11-2026
Rapidly identify and differentiate Staphylococcus aureus by detecting clumping factor, Protein A, and certain capsular polysaccharides.
Rapidly identify and differentiate Staphylococcus aureus with Thermo Scientific™ Staphytect Plus™ Latex Agglutination Test. Staphytect Plus detects clumping factor, Protein A, and certain capsular polysaccharides found in methicillin-resistant S.s aureus (MRSA) to differentiate S.s aureus from those staphylococci that do not possess these properties.
Staphytect Plus Latex Agglutination Test uses blue latex particles coated with porcine fibrinogen and rabbit IgG for detection of clumping factor, protein A and capsular polysaccharides of S. aureus.
  • Easy-to-read blue latex
  • Rapid results—within 20 seconds
  • Excellent sensitivity—detects three properties of S. aureus for wider strain identification
Traditionally, differentiation between coagulase-positive and coagulase-negative staphylococci has been performed either with the tube coagulase test that detects extracellular staphylocoagulase or the slide coagulase test that detects the clumping factor (bound coagulase) present on the bacterial cell surface. It has been reported that approximately 97% of human strains of Staphylococcus aureus possess both bound coagulase and extracelluar staphylocoagulase. Protein A is found on the cell surface of about 95% of human strains of Staphylococcus aureus and has the ability to bind the Fc portion of immunoglobulin G (IgG)1.
Certain methicillin-resistant strains of S. aureus (MRSA) may express undetectable levels of clumping factor and Protein A2,3,4 . It has been shown, however, that these strains all possess capsular polysaccharide5. The capsule can mask both Protein A and the clumping factor, thereby preventing agglutination.
Staphytect Plus uses blue latex particles coated with porcine fibrinogen and rabbit IgG, including specific polyclonal antibodies raised against capsular polysaccharides of S. aureus 6,7.
When the reagent is mixed on a card with colonies of S. aureus, rapid agglutination occurs through the reaction between (i) fibrinogen and clumping factor, (ii) the Fc portion of IgG and Protein A (iii) specific IgG and capsular polysaccharide.
Agglutination may also occur with other species which possess clumping factor or Protein A, such as Staphylococcus hyicus and Staphylococcus intermedius. If clumping factor, Protein A, or specific capsular polysaccharides are not present, agglutination will not occur and the result will be regarded as negative. The most frequent coagulase and Protein A negative isolates of staphylococci are Staphylococcus epidermidis.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2°C to 8°C
Control Sets Positive
Description Staphytect Plus
Type Staphylococcus aureus Test Kit
Includes Test latex, control latex, disposable reaction cards
Material Latex
Quantity 100 Tests/Kit
Target Organism Class Staphylococcus aureus