Learn More
Stanozolol, TRC
Product Code. 30423513
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30423513 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30423513 Supplier Toronto Research Chemicals Supplier No. TRCS684500100MG
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Stanozolol
CAS 10418-03-8
Molecular Formula C21 H32 N2 O
Quantity 100 mg
Synonym Stanozolol, 2'H-Androst-2-eno[3,2-c]pyrazol-17-ol','17-methyl-','(5α,17β)-, 2'H-5α-Androst-2-eno[3,2-c]pyrazol-17β-ol','17-methyl- (8CI), Cyclopenta[7,8]phenanthro[2,3-c]pyrazol-1-ol','1,2,3,3a,3b,4,5,5a,6,7,10,10a,10b,11,12,12a-hexadecahydro-1,10a,12a-trimethyl- (6CI,7CI), (5α,17β)-17-Methyl-2'H-androst-2-eno[3,2-c]pyrazol-17-ol, 17-Methyl-5α-androstano[3,2-c]pyrazol-17β-ol, 17-Methyl-pyrazolo[4',3':2,3]-5α-androstan-17β-ol, 17α-Methyl-17β-hydroxy-5α-androstano(3,2-c)pyrazole, 17β-Hydroxy-17-methyl-5α-androstano[3,2-c]pyrazole, Anabol, Androstanazol, Androstanazole, Androstanazolestanazol, Estazol, NSC 233046, NSC 43193, Stanabolic, Stanazolol, Stromba, Strombaject, Tevabolin, Win 14833, Winstroid, Winstrol, Winstrol Depot, Winstrol V
InChI Formula InChI=1S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24H,4-11H2,1-3H3,(H,22,23)/t14-,15+,16-,17-,19-,20-,21-/m0/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]ncc5C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name 17α-Methyl-5α-androstano[3,2-c]pyrazol-17β-ol
Molecular Weight (g/mol) 328.49
Formula Weight 328.2515