missing translation for 'onlineSavingsMsg'
Learn More

Stanozolol, TRC

Product Code. 30420407
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30420407 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30420407 Supplier Toronto Research Chemicals Supplier No. TRCS6845001G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Stanozolol
CAS 10418-03-8
Molecular Formula C21 H32 N2 O
Quantity 1 g
Synonym Stanozolol, 2'H-Androst-2-eno[3,2-c]pyrazol-17-ol','17-methyl-','(5α,17β)-, 2'H-5α-Androst-2-eno[3,2-c]pyrazol-17β-ol','17-methyl- (8CI), Cyclopenta[7,8]phenanthro[2,3-c]pyrazol-1-ol','1,2,3,3a,3b,4,5,5a,6,7,10,10a,10b,11,12,12a-hexadecahydro-1,10a,12a-trimethyl- (6CI,7CI), (5α,17β)-17-Methyl-2'H-androst-2-eno[3,2-c]pyrazol-17-ol, 17-Methyl-5α-androstano[3,2-c]pyrazol-17β-ol, 17-Methyl-pyrazolo[4',3':2,3]-5α-androstan-17β-ol, 17α-Methyl-17β-hydroxy-5α-androstano(3,2-c)pyrazole, 17β-Hydroxy-17-methyl-5α-androstano[3,2-c]pyrazole, Anabol, Androstanazol, Androstanazole, Androstanazolestanazol, Estazol, NSC 233046, NSC 43193, Stanabolic, Stanazolol, Stromba, Strombaject, Tevabolin, Win 14833, Winstroid, Winstrol, Winstrol Depot, Winstrol V
InChI Formula InChI=1S/C21H32N2O/c1-19-11-13-12-22-23-18(13)10-14(19)4-5-15-16(19)6-8-20(2)17(15)7-9-21(20,3)24/h12,14-17,24H,4-11H2,1-3H3,(H,22,23)/t14-,15+,16-,17-,19-,20-,21-/m0/s1
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4Cc5[nH]ncc5C[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name 17α-Methyl-5α-androstano[3,2-c]pyrazol-17β-ol
Molecular Weight (g/mol) 328.49
Formula Weight 328.2515
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.