Learn More
Koettermann™ Standard Table, Heavy Loads, Wheels, 1500x600x900mm
Product Code. 15313331
Change view
Click to view available options
Includes: TopResist Worktop
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15313331 Includes: TopResist Worktop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15313331 Supplier Koettermann™ Supplier No. 50700022
  • ONLINE EXCLUSIVE PRICE
    1895.00€
    1622.67€ / Pack of 1
    Save 272.33€
    14% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Koettermann™ Standard Table, Heavy Loads, Wheels, 1500x600x900mm is a mobile laboratory table with a durable worktop and is designed for heavier than normal weight bearing.
  • Heavy-duty mobile table
  • TopResist worktop
  • Standing height
  • One shelf
Specifications
Product Type Mobile Table
Depth (Metric) 750 mm
Description Heavy-Duty, Standing Height
Height (Metric) 900 mm
Includes TopResist Worktop
No. of Shelves 1
Width (Metric) 1500 mm