Learn More
Sampling Systems™ Standard Steel Sample Splitter Riffle Boxes
Product Code. 15829531 Shop All Sampling Systems Products
Click to view available options
Width (Metric) Chute:
7 mm
10 mm
15 mm
20 mm
25 mm
30 mm
40 mm
50 mm
65 mm
75 mm
80 mm
No. of Chutes:
8
10
12
Unit Size:
Each
Product Code. 15829531
Brand: Sampling Systems™ 8600M50
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These easy-to-use Sample Splitters in standard steel are for dividing and reducing bulk samples.
  • Simply drop the sample into the top and the sample is split in two
  • Split sample drops into the riffle boxes underneath
  • It is recommended that width of slots are 2.5 to 3 times bigger than the diameter of the largest particle being split
  • Available in a wide range of sizes
Specifications
Product Type Riffle Box
Material Steel
Capacity (Metric) 34.15 L
No. of Chutes 8
For Use With (Application) Dividing and Reducing Bulk Samples
Dimensions (L x W x H) 474 x 316 x 228 mm
Width (Metric) Chute 50 mm