missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 2'-OMe-iBu-G Phosphoramidite, standard grade, serum vial bottle

Product Code. 18064070 Shop All Thermo Scientific Products
Click to view available options
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 18064070

Brand: Thermo Scientific™ 27184602

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ Standard RNA Phosphoramidites ensure quality and reliability for RNA oligonucleotide synthesis.

Ensure quality and reliability for RNA oligonucleotide synthesis with Thermo Scientific™ Standard RNA 2'-TBDMS, 2'-OMe and 2'-F Phosphoramidites.

  • Rigorously controlled manufacturing processes minimize levels of reactive P(III) impurities which could be incorporated into the oligonucleotide product and/or result in the accumulation of by-products which can be difficult to remove during purification
  • NMR analysis is conducted on each lot to assure that the levels of critical impurities are within specifications

Process and Supply Chain Control:

  • Processes are strictly controlled and monitored from material procurement through final QC and product release under ISO9001 standards, providing batch-to-batch reproducibility
  • All standard RNA phosphoramidites are supplied with lot-specific quality control data including HPLC and phosphorous NMR analyses
2'-TBDMS:
  • TBDMS RNA amidite chemistry is the widely accepted standard for high quality, large-scale synthesis of RNA oligonucleotides
  • Specifications include limits for critical impurities

2'-OMe:

  • RNA oligonucleotides synthesized using 2'-OMe modified amidites are nuclease resistant, making them well suited for in vivo, antisense and aptamer applications

2'-F:

  • RNA oligonucleotides synthesized using 2'-Fluoro U and C phosphoramidites have increased thermal stability of RNA duplexes and nuclease resistance of single-stranded RNA
  • These phosphoramidites can be incorporated into custom siRNA oligonucleotides for use in gene-targeting and suppression studies

Flexible Packaging:

  • Common pack sizes fit most commercially available synthesizers and are in stock for immediate delivery
  • Custom and bulk packaging are available
TRUSTED_SUSTAINABILITY

Specifications

Format Powder
For Use With (Equipment) AKTA Synthesizers, Applied Biosystems Synthesizers
Product Type 2'-OMe-iBu-G CEP
Content And Storage Store in freezer (at or below -20°C).
Nucleoside Type Guanosine (G)
Reagent Features 2'-OMe modified
Shipping Condition Room Temperature
Purity 98%
Reagent Type Phosphoramidite
Grade Standard
Nucleic Acid Type RNA
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.