Learn More
Bochem™ Standard Retort Clamps
Product Code. p-8003051
Click to view available options
Material:
Cast Iron, Chromium-Plated
Zinc-plated Steel
Unit Size:
Each
Product Code. 10769601
Brand: Bochem™ 5464
Only null left
Add to basket
Add to basket
Corked finger. Bochem™ Standard Retort Clamps have a round jaw.
  • Round jaw
  • Cast iron chromium plated or steel zincked
  • DIN 12894
Specifications
Quantity 1 Ea.
Type Clamp
Material Zinc-plated Steel
Diameter (Metric) 80 mm
Length (Metric) Arm 180 mm
Jaw Type Round