Learn More
Tenak™ Standard Rack for Upright Freezer, 100 mm Box Height
Shop All TENAK Products
Click to view available options
Holds:
Boxes
Unit Size:
Each
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
Specifications
Specifications
| Array | 3 x 5 |
| Holds | Boxes |
| Material | Stainless Steel |
| Positions | 15 |
| For Use With (Equipment) | Upright Freezers |
| Dimensions (L x W x H) | 683 x 139 x 315 mm |
| Height (Metric) | 100 mm (Box) |