Learn More
Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 85 mm Shelf Height
Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.
Brand:  Tenak™ TE27214
Additional Details : Weight : 0.65000kg
 View more versions of this product
Product Code. 15280904
  • 83.50€ / Each
Estimated Shipment: 19-09-2024
Description
  • For use in chest freezers
  • Racks come with a locking rod to secure the cryoboxes
  • Constructed of stainless steel
Specifications
Specifications
Locking rod
Stainless Steel
135 x 92 x 437 mm
Microtiterplates (5)
Chest Frseezers
85 mm