Learn More
Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 85 mm Shelf Height
Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.
Brand: Tenak™ TE27210
Additional Details : Weight : 0.45000kg
Description
- For use in chest freezers
- Racks come with a locking rod to secure the cryoboxes
- Constructed of stainless steel
Specifications
Locking rod | |
Stainless Steel | |
135 x 92 x 264 mm |
Microtiterplates (3) | |
Chest Frseezers | |
85 mm |