Learn More
Koettermann™ Standard Hanging Cabinet, 600x780x366mm
Product Code. 15793102
Click to view available options
Locking Mechanism:
No
Yes
Door Type:
Left-Swinging Door
Right-Swinging Door
Unit Size:
Each
Product Code. 15793102
Brand: Koettermann™ 30700193
This item requires a price update prior to being ordered.
Request a Price for this item here.
Currently Unavailable: We have no information when this product will be available.
View alternative products
Koettermann™ Standard Hanging Cabinet, 600x780x366mm provides suitable storage to accommodate a variety of laboratory needs.
  • Wall-mounted cabinet
  • One door
  • One shelf
  • Side panels
Specifications
Product Type Wall-mounted Cabinet
Dimensions (D x W x H) 366 x 600 x 780 mm
Door Type Left-Swinging Door
Locking Mechanism Yes
No. of Doors 1
No. of Shelves 1