Learn More
Koettermann™ Standard Hanging Cabinet, 1500x480x350mm
Product Code. 15723092
Click to view available options
Locking Mechanism:
No
Yes
Door Type:
Open Front/No Doors
Sliding Glass Doors
No. of Doors:
2
None
Unit Size:
Each
Product Code. 15723092
Brand: Koettermann™ 30700176
This item requires a price update prior to being ordered.
Request a Price for this item here.
Currently Unavailable: We have no information when this product will be available.
View alternative products
Koettermann™ Standard Hanging Cabinet, 1500x480x350mm provides suitable storage to accommodate a variety of laboratory needs.
  • Wall-mounted cabinet
  • One shelf
  • Side panels
Specifications
Product Type Wall-mounted Cabinet
Dimensions (D x W x H) 350w, 1500 x 480 mm
Door Type Open Front/No Doors
Locking Mechanism No
No. of Doors None
No. of Shelves 1