Learn More
Corning™ Standard Conical Bioprinting Nozzles
Product Code. 17790789 Shop All Corning Products
Change view
Click to view available options
Gauge:
22 G
25 G
27 G
Nozzle Diameter (Metric):
200 μm
250 μm
410 μm
Color:
Blue
Clear
Red
Unit Size:
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Gauge Nozzle Diameter (Metric) Color Unit Size
17790789 22 G 410 μm Blue Pack of 50
17780789 25 G 250 μm Red Pack of 50
17770789 27 G 200 μm Clear Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17790789 Leverandør Corning™ Leverandørnr. 6167
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Denne vare kan ikke returneres. Se returpolicy
These Standard Conical Bioprinting Nozzles are designed fo use with Matribot™ Bioprinter. They are made from polypropylene, have a female Luer connector, and are available in 22G, 25G, and 27G.
  • Made of polypropylene
  • Feature a female Luer connector
  • Available in 22 G, 25 G, and 27 G.
TRUSTED_SUSTAINABILITY
Tekniske data
Color Blue
Product Type Bioprinting Nozzle
Material Polypropylene
Nozzle Diameter (Metric) 410 μm
Description Feature Female Luer Connector
Gauge 22 G
Length (English) 1.26 in.
Length (Metric) 32 mm
Quantity 50 Nozzles
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.