Learn More
Consort™ Conductivity Electrodes
Product Code. 11792019
Click to view available options
Temperature (Metric) Range:
0°C to 100°C
0°C to 80°C
Unit Size:
Each
Product Code. 11792019
Brand: Consort™ SK23T
Edge
Designed for laboratory, industrial, biotechnology and medical applications. Consort™ Conductivity Electrodes are high quality electrochemical analytical sensors. A range of electrodes is available for general field use, with micro volumes or for measuring high conductivities.
While calibrating or measuring, all solutions should be stirred gently to ensure a true representation of the contents. Fresh calibration solutions with values near the expected sample value should be chosen. Rinse the electrode twice between measurements: first in distilled water and then with the next sample to be measured. Allow the electrodes sufficient time to stabilize while calibrating or measuring.
  • Options include standard field model, micro-volume model and high-conductivity model
  • Built-in automatic temperature compensation (ATC Pt1000), with 2 banana connections for ATC
  • Also includes 1m cable and 1 BNC connection
  • Stability indicator prompts when readings should be taken
Specifications
Cell Constant 10 cm-1
Connector Types BNC, 2 x Banana Plug
For Use With (Application) For use with micro-volumes
Temperature (Metric) Range 0°C to 100°C
Type Conductivity Electrode
Diameter (Metric) Body 6 mm
Length (Metric) 110 mm
Length (Metric) Cable 1 m
Material Glass Body, Platinum Plate
Includes 1m cable, 2 banana connections for ATC, 1 BNC connection
Temperature Sensor Built in ATC (Pt1000)
Model SK24T