Learn More
Koettermann™ Standard Cabinet Chemical Substances, 1200x1920x516
Product Code. 14885361
Change view
Click to view available options
Dimensions (D x W x H):
516 x 1200 x 1920 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Unit Size
14885361 516 x 1200 x 1920 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14885361 Supplier Koettermann™ Supplier No. 37500007
  • ONLINE EXCLUSIVE PRICE
    2020.00€
    1901.33€ / Pack of 1
    Save 118.67€
    5% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Koettermann™ Standard Cabinet Chemical Substances, 1200x1920x516 provides suitable storage to accommodate a variety of laboratory needs, but is especially suited for storing chemicals.
  • Chemical cabinet
  • Two doors
  • Four shelves
  • Lockable
Specifications
Dimensions (D x W x H) 516 x 1200 x 1920 mm
Includes Lock
No. of Doors 2
No. of Shelves 4
Product Type Chemical Cabinet