Learn More
Koettermann™ Standard Cabinet for Acids and Bases, PP, 900x720x520
Product Code. 14805361
Change view
Click to view available options
Dimensions (D x W x H):
515 x 900 x 720 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Unit Size
14805361 515 x 900 x 720 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 14805361 Supplier Koettermann™ Supplier No. 37700011
  • ONLINE EXCLUSIVE PRICE
    4140.00€
    3901.71€ / Pack of 1
    Save 238.29€
    5% Off
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Koettermann™ Standard Cabinet for Acids and Bases, PP, 900x720x520 provides suitable storage to accommodate a variety of laboratory needs, but is especially suited for storing acids and bases.
  • Acid/base cabinet
  • Two doors
  • Three drawers
  • Fan
Specifications
Dimensions (D x W x H) 515 x 900 x 720 mm
Includes 3 Drawers, Fan
No. of Doors 2
Product Type Acids and Bases Cabinet