Learn More
Koettermann™ Standard Cabinet for Acids and Bases, PP, 520x600x1920mm
Product Code. 15518545
Change view
Click to view available options
Door Style:
Left-hinged
Right-hinged
Includes:
4 Drawers, Fan
4 Drawers, No Fan
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Door Style Includes unitSize
15518545 Right-hinged 4 Drawers, No Fan Each
14835361 Left-hinged 4 Drawers, Fan Each
14825361 Left-hinged 4 Drawers, No Fan Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15518545 Supplier Koettermann™ Supplier No. 37700002
Currently Unavailable: We have no information when this product will be available.
View alternative products
Koettermann™ Standard Cabinet for Acids and Bases provides suitable storage to accommodate a variety of laboratory needs, but is especially suited for storing acids and bases.
  • Tall, Acid/Base Cabinet
  • Two doors
  • Four drawers
Specifications
Dimensions (D x W x H) 520 x 600 x 1920 mm
Door Style Right-hinged
For Use With (Application) Acids and Bases
Includes 4 Drawers, No Fan
No. of Doors 2
Product Type Acids and Bases Cabinet