Learn More
Koettermann™ Standard Bottom Cabinet, with Wheels, 600x740x516
Product Code. 15568485
Change view
Click to view available options
No. of Drawers:
4
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Drawers Unit Size
15568485 4 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15568485 Supplier Koettermann™ Supplier No. 31500076
  • ONLINE EXCLUSIVE PRICE
    1330.00€
    1079.03€ / Pack of 1
    Save 250.97€
    18% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Koettermann™ Standard Bottom Cabinet, with Wheels, 600x740x516 provides suitable storage to accommodate a variety of laboratory needs.
  • Mobile cabinet
  • Four drawers
  • Lockable
Specifications
Product Type Mobile Storage Cabinet
Dimensions (D x W x H) 516 x 600 x 740 mm
Locking Mechanism Yes
No. of Drawers 4