Learn More
Rogo Sampaic™ Stand
Product Code. p-8008549
Click to view available options
Size:
180 mm
120 x 210 mm
Unit Size:
Each
Product Code. 11702674
Brand: Rogo Sampaic™ FFB515
Only null left
Specifications
Product Type Stand
Length (Metric) 210 mm
Size 120 x 210 mm
Dimensions (L x W) 210 x 120 mm
Height (English) 3 ft.
No. per Pack 1 Ea.
Width (Metric) 120 mm