Learn More
Schuett Biotec™ Stainless Steel Turnable
Click to view available options
Diameter (Metric):
160 mm
Height (Metric):
45 mm
70 mm
Type:
Turnable
Unit Size:
Each
Specifications
Specifications
| Material | Stainless Steel |
| Type | Turnable |
| For Use With (Equipment) | Petri Dishes Petriturn-Mplus |
| Height (Metric) | 45 mm |
| Diameter (Metric) | 160 mm |