Learn More
Lauda™ Stainless Steel Tubing Connectors
Lauda™ Stainless Steel Tubing Connectors are used for connecting polymer tubing to LAUDA thermostats and circulation chillers.
Brand: Lauda™ HKO 025
Additional Details : Weight : 30.00000kg
Description
- For connecting polymer tubings
Specifications
Tubing Connector | |
Stainless Steel | |
11 mm |