Learn More
JP Selecta™ Stainless Steel Tripod Support Stand
Click to view available options
Diameter (Metric) Outer:
8 mm
12 mm
Height (Metric):
500 mm
700 mm
Weight (Metric):
1.4 kg
2.5 kg
Unit Size:
Each
Description
- Made of AISI 304 stainless steel
Specifications
Specifications
| Material | Stainless Steel |
| Height (Metric) | 700 mm |
| Diameter (Metric) Outer | 12 mm |
| Product Type | Tripod support rod |
| Quantity | 1 Ea. |
| Weight (Metric) | 2.5 kg |