Learn More
Amsta Rostfritt Stainless Steel Transport Container Without Skirtband, Without Lid
Click to view available options
Capacity (Metric):
20 L
25 L
30 L
50 L
75 L
100 L
Unit Size:
Each
Description
- Stainless steel 304 /1.4301
- Straight cylindrical vessel in highly polished finish with stylish design.
Specifications
Specifications
| Product Type | Pail |
| Capacity (Metric) | 50 L |
| Color | Silver |
| Material | Stainless Steel |