Learn More
Sampling Systems™ Stainless Steel Test Tube Rack
Shop All Sampling Systems Products
Click to view available options
Positions:
12 Positions
48 Positions
Array:
2 x 6
4 x 12
Unit Size:
Each
Description
- Made from corrosion resistant 304 stainless steel
- Ideal for use in laboratories and production
- Mirror polish finish
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Silver |
| Holds | Test Tubes |
| Material | Stainless Steel |
| Positions | 48 Positions |
| Shape | Rectangular |
| Stackable | Yes, When Empty |
| Sterility | Non-sterile |
| Product Type | Test Tube Rack |
| Shelves | None |
| Show More |