Learn More
Grant Instruments™ Stainless Steel Test Tube Rack
Product Code. 15385842
Click to view available options
For Use With (Equipment):
0.5mm dia. Tubes
1.5mm dia. Tubes
10mm dia. Tubes
13mm dia. Tubes
16mm dia. Tubes
19mm dia. Tubes
25mm dia. Tubes
30mm dia. Tubes
Unit Size:
Each
Product Code. 15385842
Brand: Grant Instruments™ SRSE
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Fits a variety of test tube sizes from 10 to 30mm diameter
  • For use with LSB Aqua Pro
  • Available in 8 sizes to hold up to 48 SR test tubes
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 119 mL
For Use With (Equipment) 0.5mm dia. Tubes
Material Stainless Steel