Learn More
RETSCH Stainless-Steel Test Sieves, 450 mm Dia., in Pore Sizes 10 mm and Smaller with ISO 3310/1
Product Code. p-8000560 Shop All Retsch Products
Click to view available options
Pore Size:
1 mm
1.12 mm
1.18 mm
1.25 mm
1.4 mm
1.6 mm
1.7 mm
1.8 mm
2 mm
2.24 mm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
3.35 mm
3.55 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
6.7 mm
7.1 mm
8 mm
9 mm
9.5 mm
10 mm
Unit Size:
Each
Product Code. 11984572
Brand: RETSCH 60.168.001120
Only null left
Specifications
Quantity 1 Each
Pore Size 1.12 mm
Type Test Sieve
Certifications/Compliance ISO 3310/1
For Use With (Equipment) Vibratory Shaker AS 450
Includes Certificate indicating Compliance
Height (Metric) 100mm
For Use With (Application) For separation, fractioning, particle size determination
Diameter (Metric) Outer 450 mm
Frame Material Stainless Steel
Mesh Material Stainless Steel