Learn More
Linker Industrietechnik™ Stainless Steel Test Sieves
Click to view available options
Pore Size:
63 μm
1 mm
150 μm
200 μm
250 μm
400 μm
500 μm
800 μm
Unit Size:
Each
Description
- Stainless steel ring construction
- Stainless steel sieve cloth/skin
- Soft solder seam
- Heat resistant
- Available in a variety of sieve mesh sizes
Specifications
Specifications
| Description | Heat resistant up to 170°C |
| Pore Size | 250 μm |
| Certifications/Compliance | 2.1 EN 10 204, ISO 3310-1 |
| Height (Metric) | Internal: 25 mm |
| U.S. Standard | No. 60 |
| Diameter (Metric) Outer | 50 mm |
| Frame Material | Stainless Steel |
| Screen Size | 60 Mesh |
| Mesh Material | Stainless Steel |
| Sieve Number | 60 |