Learn More
Linker Industrietechnik™ Stainless Steel Test Sieves
Product Code. 11799249
Click to view available options
Pore Size:
63 μm
1 mm
150 μm
200 μm
250 μm
400 μm
500 μm
800 μm
Unit Size:
Each
Product Code. 11799249
Brand: Linker Industrietechnik™ 552.025.222035
Constructed with a soft solder seam, and a sieve of stainless steel cloth.
  • Stainless steel ring construction
  • Stainless steel sieve cloth/skin
  • Soft solder seam
  • Heat resistant
  • Available in a variety of sieve mesh sizes
Specifications
Description Heat resistant up to 170°C
Pore Size 200 μm
Certifications/Compliance 2.1 EN 10 204, ISO 3310-1
Height (Metric) Internal: 25 mm
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Screen Size 200 μm
Mesh Material Stainless Steel