Learn More
Swann-Morton™ Stainless Steel Surgical Scalpels
Click to view available options
Size:
# 10A
# 12
# 15
# 20
# 21
# 22
# 22A
# 23
# 24
Unit Size:
Pack of 10
Description
- Stainless steel blade, curved
- Polystyrene handle
- Individually wrapped
- Sterile
- Disposable
Specifications
Specifications
| Blade Material | Stainless Steel |
| For Use With (Application) | For Arteriotomies, Parotid Surgeries, Mucosal Cuts on a Septoplasty and during Cleft Palate Procedures, Ureterolithotomies |
| Size | # 12 |
| Description | Scalpel, Curved Blade |
| Disposable | Single-use |
| Sterility | Sterile |
| Handle Material | Polystyrene |