Learn More
Fisherbrand™ Stainless Steel Stirring Shafts
Shop All Fisherbrand Products
Click to view available options
Includes:
Anchor blade
Paddle blade, 6 holes
Weight (Metric):
0.01 kg
0.99 kg
Unit Size:
Each
Specifications
Specifications
| For Use With (Equipment) | Overhead Stirrer |
| Includes | Anchor blade |
| Quantity | 1 Each |
| Type | Stirring Shaft |
| Weight (Metric) | 0.01 kg |
| Material | Stainless Steel |