Learn More
Linker Industrietechnik™ Stainless Steel Lid for 200mm dia.Sieves
Product Code. 11734429
Click to view available options
Diameter (Metric) Outer:
200 mm
Unit Size:
Each
Product Code. 11734429
Brand: Linker Industrietechnik™ 552.025.200901
Only null left
Add to basket
Add to basket
Cover sieves to avoid loss of sample during separation process. This Stainless Steel lid will fit 200 mm diameter sieves, and is recommended for sieves with 25 mm height.
Stainless Steel lid can be used with 200mm diameter test sieves that have an internal height of 25mm.
Specifications
Quantity 1 Ea.
Type Sieve Cover
Material Stainless Steel
For Use With (Application) For 200 dia. x 25mm internal height sieves
Diameter (Metric) Outer 200 mm