Learn More
Memmert Stainless steel slide-in drip tray, 15mm (for reinforced chamber option), for ICH750
Brand:  Memmert ICH750B32763
Stainless steel slide-in drip tray, 15mm (for reinforced chamber option), for ICH750
Product Code. 16356687
  • ONLINE EXCLUSIVE PRICE
    315.00€
    286.00€ / Each
    Save 29.00€
    9% Off
Only null left
Estimated Shipment: 16-03-2026