Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Product Code. p-8013613 Shop All Kinematica Products
Click to view available options
Pore Size:
0.2 mm
0.5 mm
0.8 mm
1 mm
1.5 mm
2.0 mm
3 mm
4 mm
5 mm
6 mm
Unit Size:
Each
Product Code. 10012393
Brand: Kinematica™ 35095008
Depending on the sample, even finer results can be achieved by grinding with two or more sieves with progressively smaller hole sizes.
Specifications
Product Type Sieve
For Use With (Equipment) PX MFC 90 D
Material Stainless Steel
Pore Size 0.8 mm