Learn More
Kinematica™ Stainless Steel Sieve for Polymix™ MFC 90 D Mill
Shop All Kinematica Products
Click to view available options
Pore Size:
0.2 mm
0.5 mm
0.8 mm
1 mm
1.5 mm
2.0 mm
3 mm
4 mm
5 mm
6 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Sieve |
| For Use With (Equipment) | PX MFC 90 D |
| Material | Stainless Steel |
| Pore Size | 6 mm |