Learn More
Linker Industrietechnik™ Stainless Steel Sieve Bottom without Drain
Click to view available options
Diameter (Metric) Outer:
200 mm
Unit Size:
Each
Description
- Square mesh
- Brushed Stainless Steel frame
- Maximum temperature of 170°C
Specifications
Specifications
| Quantity | 1 Each |
| Type | Sieve Bottom |
| Material | Stainless Steel |
| Height (Metric) | 50 mm |
| Diameter (Metric) Outer | 200 mm |