Learn More
RETSCH Stainless-Steel Seven Analysis Test Sieves, 200mm Dia.
Brand:  RETSCH 60.322.000630
171.95 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10370432
  • 181.00€ / Each
Estimated Shipment: 10-07-2024
Specifications
Stainless Steel
Stainless Steel
1 Each
Test Sieve
25mm
Separation, fractioning, particle size determination
200 mm
630μm
ISO 3310 and ASTM E 11