Learn More
De Buyer™ Stainless Steel Serving Dishes
Product Code. 11900625
Click to view available options
Volume (Metric):
3.6 L
4.9 L
Unit Size:
Each
Product Code. 11900625
Brand: De Buyer™ 3280.40
Only null left
Specifications
Product Type Tray
Material Stainless Steel
Height (Metric) 50 mm
Length (Metric) 420 mm
Quantity Each
Volume (Metric) 4.9 L
Width (Metric) 320 mm