Learn More
Rogo Sampaic™ Stainless Steel Scissors
Click to view available options
Length (Metric):
110 mm
115 mm
120 mm
140 mm
Tip Style:
Round/Round
Sharp/Round
Sharp/Sharp
For Use With (Application):
Anatomical
Dissecting
Iris
Metzenbaum
Noyes micro
Unit Size:
Each
Pack of 10
Description
- Surgical grade stainless steel
- Available in a variety of styles - Metzenbaum, dissecting, iris, anatomical
Specifications
Specifications
| Product Type | Stainless Steel Scissors |
| Length (Metric) | 110 mm |
| Material | Stainless Steel |
| For Use With (Application) | Iris |
| Tip Style | Sharp/Sharp |