Learn More
Rogo Sampaic™ Stainless Steel Scalpel
Product Code. 11718551
Change view
Click to view available options
Length (Metric) Blade:
50 mm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Blade Unit Size
11718551 50 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11718551 Supplier Rogo Sampaic™ Supplier No. FMB025
  • ONLINE EXCLUSIVE PRICE
    26.80€
    23.04€ / Pack of 10
    Save 3.76€
    14% Off
Estimated Shipment: 21-10-2026
Order Info
LengthMetricBlade
Specifications
Blade Material Stainless Steel
Product Type Scalpel
Length (Metric) Blade 50 mm