Learn More
Rotronic™ Stainless Steel Sample Holder For Water Activity Probes HC2-AW(-USB)
Product Code. 11900685
Change view
Click to view available options
Product Type:
Sample Holder
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
11900685 Sample Holder Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11900685 Supplier Rotronic™ Supplier No. WP40
  • ONLINE EXCLUSIVE PRICE
    731.57€
    533.00€ / Pack of 1
    Save 198.57€
    27% Off
Estimated Shipment: 20-11-2026
Specifications
Product Type Sample Holder
For Use With (Equipment) For Water Activity Probes HC2-AW(-USB)
Material Stainless Steel
Quantity 1 Each