Learn More
Aalborg™ Stainless Steel Utility Needle Valve
Product Code. 11727549
Click to view available options
Airflow Rate:
20000 mL/min.
Unit Size:
Each
Product Code. 11727549
Brand: Aalborg™ VCMSV2A
Only null left
Add to basket
Add to basket
These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes. Aalborg™ Stainless Steel Utility Needle Valve is designed for controlling a broad range of flow rates of liquids and gases.
  • Available in three conveniently overlapping orifice-needle sizes.
  • These versatile, rugged and reliable, valves are suitable for laboratory instrumentation, bench top or OEM flow control purposes.
  • Valves are offered in straight (T) flow patterns. All valves are supplied with 1/8in. FNPT inlet and outlet ports.
  • Valve cartridges are also interchangeable with built-in valves of Aalborg’s series of P, T, S, and G flow meter product line.
  • Bubble tight shutoff
Specifications
Type Right Standard Valve
Airflow Rate 20000 mL/min.
Waterflow Rate 1200mL/min.
Max. Operating Temperature (Metric) 121°C