Learn More
Thermo Scientific™ Stainless Steel Reinforced Shelf
Product Code. 15726064 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
14 kg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15726064 14 kg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15726064 Supplier Thermo Scientific™ Supplier No. 50150643
  • ONLINE EXCLUSIVE PRICE
    423.00€
    398.00€ / Pack of 1
    Save 25.00€
    5% Off
View all promotions
 Restricted Item
In Stock
Stainless Steel Reinforced Shelf
  • Stainless Steel Reinforced Shelf
  • For Heracell VIOS 250/240i CO2 Incubator
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 14 kg
CO2 Sensor Technology No Sensor
For Use With (Equipment) Heracell VIOS 250/240 i CO2 Incubator
Lighting No Lighting
Material Stainless Steel
Oxygen Control None
Relative Humidity None
Type Reinforced Shelf