Learn More
Agencinox™ Stainless Steel Rectangular Tray
Click to view available options
Dimensions (L x W x H):
220 x 145 x 28 mm
245 x 175 x 30 mm
265 x 185 x 30 mm
285 x 185 x 30 mm
300 x 210 x 40 mm
355 x 255 x 40 mm
425 x 280 x 50 mm
510 x 340 x 50 mm
Unit Size:
Each
Description
- Rectangular tray
- Stainless steel construction
- Available in a variety of sizes
Specifications
Specifications
| Dimensions (L x W) | 425 x 280 mm |
| Dimensions (L x W x H) | 425 x 280 x 50 mm |
| Height (Metric) | 50 mm |
| Length (Metric) | 425 mm |
| Material | Stainless Steel |
| Type | Tray |
| Width (Metric) | 280 mm |
| For Use With | Instruments |
| Shape | Rectangular |