Learn More
Bochem™ Stainless Steel Rectangle Test Tube Stands
Click to view available options
Positions:
12
24
48
Array:
2 x 12
2 x 6
4 x 12
Diameter (Metric) Opening:
17 mm
20 mm
Unit Size:
Each
Specifications
Specifications
| Product Type | Test Tube Rack |
| Array | 2 x 12 |
| Material | Stainless Steel (18/10) |
| Positions | 24 |
| Diameter (Metric) Opening | 20 mm |