Learn More
Cole-Parmer™ Straight Fitting, Quick-Disconnect, Push-to-Connect x NPT(M), Stainless Steel
Click to view available options
:
Fits tubing accepting 1/4 in. push-to-connect and 1/8 in. NPT
Description
- Durable stainless steel construction makes these fittings ideal for harsh environments that demand chemically resistant fluid lines.
- Whether required for a process area with frequent detergent washdowns or exposure to highly corrosive chemicals, these fittings ensure uninterrupted air or fluid lines.
- Matching material of construction to your stainless piping will reduce the effects of thermal expansion differentials and minimize line leakage.
Specifications
Specifications
| Material | Stainless Steel |
| For Use With | 1/4 in. push-to-connect female insert and 1/8 in. NPT(F) connection |
| Min. Operating Temperature (Metric) | -20°C |
| Packaging Quantity | 1 Pack |
| Tubing Size | Fits tubing accepting 1/4 in. push-to-connect and 1/8 in. NPT |
| Material (Seal) | FKM |
| Maximum Pressure | 225 psi |
| Vacuum Level | Max: 762 mm Hg |
| Max. Operating Temperature (Metric) | 149°C |
| Product Type | Push-to-connect Male Threaded Adapter |