Learn More
Linker Industrietechnik™ Stainless Steel Test Sieve with Soft Soldered Seam, 200 Dia. x 50 mm Height
Product Code. 11746737
Click to view available options
Pore Size:
40 μm
63 μm
100 μm
200 μm
2.36 mm
2.5 mm
2.8 mm
3.15 mm
315 μm
3.35 mm
4 mm
4.5 mm
4.75 mm
5 mm
5.6 mm
6.3 mm
7.1 mm
8 mm
9 mm
10 mm
Unit Size:
Each
Product Code. 11746737
Brand: Linker Industrietechnik™ 552.050.222075
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Sifts particles in a wide range of pore sizes. These stainless-steel test sieves have soft soldered seam and an internal height of 50 mm and diameter of 200 mm.
  • Stainless-steel racks with an internal height of 50 mm
  • Diameter: 200 mm
  • Stainless-steel test sieve skin
  • Soft soldered seam
  • Identification number and certificate of compliance
  • Resistant up to temperatures of 170°C
Specifications
Quantity 1Each
Pore Size 6.3 mm
Type Test Sieve
Certifications/Compliance Compliance 2.1 EN 10 204 and ISO 3310-1
Material Stainless Steel
Height (Metric) 50mm
Diameter (Metric) Outer 200 mm
Mesh Material Stainless Steel