Learn More
RETSCH Stainless Steel Push-fit Feed Chutes
Product Code. p-8004981 Shop All Retsch Products
Click to view available options
Dimensions (L x W):
210 x 15 mm
Unit Size:
Each
Product Code. 10064721
Brand: RETSCH 03.729.0035
Only null left
Specifications
Product Type Push-fit Feed Chute
Dimensions (L x W) 210 x 15 mm
For Use With (Equipment) For Holder 15/40 DR 100
Length (Metric) 210 mm
Material Stainless Steel
Width (Metric) 15 mm