Learn More
RUBIS High Precision Tweezers - SwissTweeze
Product Code. 11769564
Encompass_Preferred
missing translation for 'orderingAttributeHoverText'
Length (Metric):
110 mm
115 mm
120 mm
Tip Style:
Needle Pointed, Ultra Fine
Pointed, Fine
Pointed, Ultra Fine
Round, Flat, Fine
Design:
Angled
Angled, Bent
Curved
Straight
missing translation for 'unitSize'
Each
Product Code. 11769564
missing translation for 'mfr': RUBIS 2AG
Only null left
Add to basket
Add to basket
High Precision Tweezers - Antimagnetic/Anti-acid SS, Perforated Handle
  • High Precision Tweezers
  • 100% Made in Switzerland
  • Anti-acid/Antimagnetic Stainless Steel
  • Perfect alignment
  • Very effective
  • Reliable
  • Excellent chemical resistance
  • Swiss Tweeze series
  • Perforated handle for enhanced grip
Specifications
Tip Style Round, Flat, Fine
Design Straight
Length (English) 4.52 in.
Material Anti-acid, Anti-magnetic Stainless Steel
Length (Metric) 110 mm
Product Type High Precision Tweezers
Quantity (No. per Case) Each
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.